missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD49c (Integrin alpha 3) Monoclonal Antibody (5F1D4), Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen MA548513
This item is not returnable.
View return policy
Description
ITGA3 (INTEGRIN, ALPHA-3), also called CD49C, VLA3 or GAPB3, is a protein that in humans is encoded by the ITGA3 gene. ITGA3 is an integrin alpha subunit which is also a member of the family of cell surface adhesion molecules. ITGA3 is mapped to chromosome 17 and its exact cytogenetic location is 17q21.33. ITGA3 makes up half of the alpha3beta1 integrin duplex that plays a role in neural migration and corticogenesis together with beta-1 subunit. A functional link between DAB1 phosphorylation and ITGA3 signaling drives the timely detachment of migrating neurons from radial glial fibers. Expression of human ITGA3 increased the infectivity of virus for Chinese hamster ovary cells. ITGA3 also contains 13 potential N-glycosylation sites, 2 potential cleavage sites, and the 7 N-terminal repeating units characteristic of ITGAs. Recombinant ITGA3 is expressed as a 150-kD protein as the same size as the native protein by the western blot analysis.
Specifications
CD49c (Integrin alpha 3) | |
Monoclonal | |
1 mg/mL | |
PBS with 0.05% sodium azide | |
P26006 | |
ITGA3 | |
Purified recombinant fragment of human CD49C (AA: extra 63-248) expressed in E. Coli. | |
100 μg | |
Primary | |
Human | |
Liquid |
ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Western Blot | |
5F1D4 | |
Unconjugated | |
ITGA3 | |
AA407068; alpha 3 subunit of VLA-3 receptor; Alpha3 integrin; antigen CD49C; antigen identified by monoclonal antibody J143; CD49 antigen-like family member C; CD49c; CD49c antigen; FLJ34631; FLJ34704; FRP-2; Galactoprotein B3; GAPB3; GAP-B3; ILNEB; integrin alpha 3; Integrin alpha3; Integrin alpha-3; Integrin alpha-3 heavy chain; Integrin alpha-3 light chain; integrin subunit alpha 3; integrin, alpha 3; integrin, alpha 3 (antigen CD49C, alpha 3 subunit of VLA-3 receptor); ITGA3; MSK18; testicular tissue protein Li 96; testicular tissue protein Li 98; VCA-2; very late activation protein 3 receptor, alpha-3 subunit; VL3A; VLA-3 subunit alpha; VLA3a | |
Mouse | |
Protein G | |
RUO | |
3675 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CD49c (Integrin alpha 3) Monoclonal Antibody (5F1D4), Invitrogen™ > 100 μg; Unconjugated
Spot an opportunity for improvement?Share a Content Correction