Learn More
CD4 Mouse, anti-Human, FITC, Clone: RPA-T4, STEMCELL Technologies
Mouse Monoclonal Antibody
Marca: Stemcell Technologies 100-0302
Descripción
- The RPA-T4 antibody reacts with CD4, an ∼59 kDa single-chain type 1 transmembrane glycoprotein and member of the immunoglobulin (Ig) superfamily; CD4 contains four extracellular Ig-like domains (D1 - D4)
- The epitope for RPA-T4 has been localized to the D1 domain of the protein, which has a structure resembling an Ig variable domain
- CD4 is expressed at relatively high levels by most thymocytes and a subpopulation of T cells (T helper cells), and at lower levels by peripheral blood monocytes and macrophages
- CD4 binds to a nonpolymorphic region of MHC II and acts as a co-receptor to the T cell receptor (TCR) in MHC II-restricted antigen recognition by enhancing the affinity of the association between the TCR and MHC II-antigen complex
- CD4 also functions to amplify signals from the TCR to the cytoplasm through the interaction of its intracellular domain with cytoplasmic tyrosine kinases, such as Lck
- Moreover, CD4 is a receptor for the human immunodeficiency virus (HIV)
- Binding of the RPA-T4 antibody blocks HIV binding and mixed lymphocyte reaction (MLR), and inhibits the activation of CD4+ T cells in vitro by interfering with the association of CD4 and HLA-DR
- It also partially blocks the binding of antibody clone SK3
- The epitope of the RPA-T4 antibody is reportedly sensitive to formaldehyde fixation
Especificaciones
CD4 | |
Monoclonal | |
FITC | |
100 Tests | |
Chimpanzee, Human |
CyTOF, Western Blot, Immunocytochemistry, Immunofluorescence, Functional Assay | |
RPA-T4 | |
Mouse | |
Primary | |
T Cells |
El Programa Encompass de Fisher Scientific ofrece artículos que no forman parte de nuestra cartera de distribución. Estos productos no suelen tener imágenes ni descripciones detalladas. Sin embargo, nos comprometemos a mejorar su experiencia de compra. Utilice el siguiente formulario para hacernos llegar sus comentarios sobre el contenido de este producto.