Learn More
CD33 Mouse, anti-Human, PE, Clone: P67.6, STEMCELL Technologies
Mouse Monoclonal Antibody
Marca:  Stemcell Technologies 60126PE
Encompass_Preferred
 Ver más versiones de este producto
Código de producto. 17109161
  • 529.20€ / 100 tests
Descripción
  • The P67.6 antibody reacts with CD33 (Siglec-3), a 67 kDa type 1 transmembrane sialoadhesin protein of the immunoglobulin superfamily
  • CD33 is expressed on committed myeloid progenitor cells, monocytes, granulocytes, dendritic cells, mast cells, resident macrophages, subsets of B cells, and activated T and NK cells
  • CD33 is a major marker used to differentiate myeloid from lymphoid or erythroid leukemias
  • In peripheral blood, it is expressed at high levels on monocytes and at lower levels on granulocytes
  • CD33 functions as a sialic acid-dependent cell adhesion molecule and mediates protein-glycan and protein-protein interactions between cells
  • It also has a putative role as an inhibitory receptor during the immune response
Especificaciones
Especificaciones
CD33
Monoclonal
PE
100 Tests
Human
Granulocytes and Subsets, Myeloid Cells
Flow Cytometry
P67.6
Mouse
Primary
IgG1 κ
Certificados
Promociones
Promociones
El Programa Encompass de Fisher Scientific ofrece artículos que no forman parte de nuestra cartera de distribución. Estos productos no suelen tener imágenes ni descripciones detalladas. Sin embargo, nos comprometemos a mejorar su experiencia de compra. Utilice el siguiente formulario para hacernos llegar sus comentarios sobre el contenido de este producto.
Título del producto
CD33 Mouse, anti-Human, PE, Clone: P67.6, STEMCELL Technologies