Learn More
CD33 Chimeric Recombinant Rabbit Monoclonal Antibody (M195), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Marca: Invitrogen MA548347
Descripción
Specificity: This antibody binds CD33, which is a 67 kD glycoprotein expressed on monocytes, hematopoetic progenitors, early myeloid progenitor cells and myeloid leukemias (acute and chronic) but not expressed on mature myeloid cells, normal stem cells, other hemtopoietic cells and non hematopoietic human tissues.
CD33 is a transmembrane protein of the sialic acid-binding immunoglobulin-like lectin (Siglec) family. It belongs to the immunoreceptor tyrosine-based inhibitory motif (ITIM)-containing molecules able of recruiting protein tyrosine phosphatases SHP-1 and SHP-2 to signal assemblies, and these ITIMs are also used for ubiquitin-mediated removal of the receptor from the cell surface. CD33 is expressed on cells of myelomonocytic lineage, binds sialic acid residues in N- and O-glycans on cell surfaces, and is a therapeutic target for acute myeloid leukemia. Further, CD33 is found on granulocyte and macrophage precursors in the bone marrow, but is not on pluripotent stem cells. CD33 is also expressed on, and is a useful marker for, peripheral monocytes. CD33 is useful for distinguishing myelogenous leukemia cells from lymphoid or erythroid leukemias. Diseases associated with CD43 dysfunction include gallbladder lymphoma and extracutaneous mastocytoma.
Especificaciones
CD33 Chimeric | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with 0.02% ProClin 300 | |
P20138 | |
CD33 | |
The original antibody was generated by immunizing mice with live leukemic myeloblasts. | |
200 μg | |
Primary | |
Human | |
Liquid |
Flow Cytometry, Functional Assay, Immunocytochemistry, Radioimmune Assays (RIA) | |
M195 | |
Unconjugated | |
CD33 | |
CD33; CD33 antigen; CD33 antigen (gp67); CD33 molecule; CD33 molecule transcript; FLJ00391; gp67; Myeloid cell surface antigen CD33; p67; sialic acid binding Ig-like lectin 3; sialic acid-binding Ig-like lectin 3; SIGLEC3; Siglec-3 | |
Rabbit | |
Affinity chromatography | |
RUO | |
945 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
IgG κ |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.