Learn More
CD279 (PD-1) Mouse anti-Human, APC, Clone: J110, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech APC-65119-100TESTS
Descripción
The CD279 (PD-1) antibody from Proteintech is a mouse monoclonal antibody generated with n/a of human CD279 (PD-1). This antibody recognizes human antigen. The CD279 (PD-1) antibody has been validated for the following applications: FC analysis. This antibody is apc.
Especificaciones
CD279 (PD-1) | |
Monoclonal | |
5 μL (1 μg)/Test | |
Mouse | |
Primary | |
Human |
Flow Cytometry | |
J110 | |
APC | |
100 Tests | |
5133 | |
IgG1 |