Learn More
CD19 Rat, anti-Mouse, Biotin, Clone: 6D5, STEMCELL Technologies
Rat Monoclonal Antibody
Marca: Stemcell Technologies 60006BT.1
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
- The 6D5 antibody reacts with CD19, an ∼95 kDa type 1 transmembrane glycoprotein expressed on the surface of B cells throughout all stages of development, from early pre-B cells to plasma cells
- Expression is down-regulated but persists in terminally differentiated plasma cells
- CD19 is also found on follicular dendritic cells
- By associating with CD21 and CD81, CD19 functions as a co-receptor for the B cell receptor and is involved in B cell activation and differentiation
- Activation of CD19 is accompanied by phosphorylation of the cytoplasmic domain, which promotes binding to kinases and the induction of intracellular signaling cascades
- Mutations in CD19 can result in severe immunodeficiency syndromes
- Clone 6D5 recognizes the same epitope as clone ID3 in cross-competition assays
- This antibody clone has been verified for purity assessments of cells isolated with EasySep™ kits, including EasySep™ Mouse B Cell Isolation Kit (Catalog #19854)
Especificaciones
| CD19 | |
| Monoclonal | |
| Biotin | |
| 50 μg | |
| Mouse | |
| B Cells |
| Immunohistochemistry, Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| 6D5 | |
| Rat | |
| Primary | |
| IgG2a κ |