Learn More
CD127 (IL-7Ra) Rat anti-Mouse, PE, Clone: A7R34, Proteintech
Rat Monoclonal Antibody
Marca:  Proteintech PE-65093-100UG
Código de producto. 16851696
  • 104.00€ / 100 microgramos
Descripción
The CD127 (IL-7Ra) antibody from Proteintech is a rat monoclonal antibody generated with il-7ra-igg1 fusion protein of mouse CD127 (IL-7Ra). This antibody recognizes mouse antigen. The CD127 (IL-7Ra) antibody has been validated for the following applications: FC analysis. This antibody is pe.
Especificaciones
Especificaciones
CD127 (IL-7Ra)
Monoclonal
0.2 mg/mL
Rat
Primary
Mouse
Flow Cytometry
A7R34
PE
100 μg
16197
IgG2a
Documentos
One moment while we fetch your results.
Certificados
Promociones
Promociones
Título del producto
CD127 (IL-7Ra) Rat anti-Mouse, PE, Clone: A7R34, Proteintech > 100 μg