Learn More
CD11b, APC, clone: VIM12, Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen CD11B05
Descripción
CD11b is implicated in various adhesive interactions of monocytes, macrophages and granulocytes as well as in mediating the uptake of complement coated particles. It is identical to CR3, the receptor for the iC3b fragment of the third complement component. It probably recognizes the RGD peptide in C3b. CD11b is also a receptor for fibrinogen, factor X, and ICAM1. It recognizes P1 and P2 peptides of fibrinogen gamma chain. The Mac1 CD11b antigen is present on macrophages, granulocytes, natural killer cells, and blood monocytes. CD11b is expressed on 8% of spleen cells, 44% of bone marrow cells, and less than 1% of thymocytes, and is commonly used as a microglial marker in nervous tissue.
Allophycocyanin (APC) is a stable and highly soluble phycobiliprotein that provides maximal absorbance and fluorescence without susceptibility to internal or external fluorescence quenching, thus providing exceptional quantum yields and molar extinction coefficients.
Especificaciones
CD11b | |
Monoclonal | |
APC | |
ITGAM | |
CD11B, CR3A, MAC-1, MAC1A, MGC117044, MO1A, SLEB6, Integrin alpha M | |
Mouse | |
Purified | |
ASR, ASR/IVD | |
3684 | |
Store at 2 - 8 °C. | |
Liquid |
Flow Cytometry | |
VIM12 | |
PBS with BSA and 0.1% sodium azide | |
P11215 | |
ITGAM | |
Human CD11b (Mac-1) | |
0.5 mL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.