Learn More
CD105 (Endoglin) Mouse, anti-Human, PE, Clone: 43A3, STEMCELL Technologies
Mouse Monoclonal Antibody
Marca: Stemcell Technologies 60039PE
Descripción
- The 43A3 antibody reacts with CD105 (endoglin), an ∼180 kDa cell surface glycoprotein which is a disulfide-bonded homodimer of ∼90 kDa type I transmembrane subunits
- CD105 is a component of the TGF-β receptor complex and is expressed by vascular endothelial smooth muscle cells, syncytiotrophoblasts of placenta and activated macrophages, and at relatively low levels by stromal fibroblasts
- Its expression is also observed in some types of tumors, and levels are up-regulated on the endothelium during angiogenesis
- CD105 has important roles in angiogenesis, cardiovascular development, and vascular remodeling, and the protein serves a regulatory role in cytoskeletal reorganization by modulating the sites of focal adhesion and cellular migration
- This antibody clone has been verified for labeling human mesenchymal cells grown in MesenCult™ Proliferation Kit (Human; Catalog #05411) and MesenCult™-XF Medium (Catalog #05420)
Especificaciones
CD105 (Endoglin) | |
Monoclonal | |
PE | |
100 Tests | |
Human, Mouse | |
Hematopoietic Stem and Progenitor Cells, Mesenchymal Stem and Progenitor Cells |
Immunoprecipitation, Western Blot, Flow Cytometry | |
43A3 | |
Mouse | |
Primary | |
IgG1 κ |
El Programa Encompass de Fisher Scientific ofrece artículos que no forman parte de nuestra cartera de distribución. Estos productos no suelen tener imágenes ni descripciones detalladas. Sin embargo, nos comprometemos a mejorar su experiencia de compra. Utilice el siguiente formulario para hacernos llegar sus comentarios sobre el contenido de este producto.