Learn More
CD105 (Endoglin) Mouse, anti-Human, PE, Clone: 43A3, STEMCELL Technologies
Mouse Monoclonal Antibody
Marca:  Stemcell Technologies 60039PE
Encompass_Preferred
 Ver más versiones de este producto
Código de producto. 17188711
  • 495.45€ / 100 tests
Descripción
  • The 43A3 antibody reacts with CD105 (endoglin), an ∼180 kDa cell surface glycoprotein which is a disulfide-bonded homodimer of ∼90 kDa type I transmembrane subunits
  • CD105 is a component of the TGF-β receptor complex and is expressed by vascular endothelial smooth muscle cells, syncytiotrophoblasts of placenta and activated macrophages, and at relatively low levels by stromal fibroblasts
  • Its expression is also observed in some types of tumors, and levels are up-regulated on the endothelium during angiogenesis
  • CD105 has important roles in angiogenesis, cardiovascular development, and vascular remodeling, and the protein serves a regulatory role in cytoskeletal reorganization by modulating the sites of focal adhesion and cellular migration
  • This antibody clone has been verified for labeling human mesenchymal cells grown in MesenCult™ Proliferation Kit (Human; Catalog #05411) and MesenCult™-XF Medium (Catalog #05420)
Especificaciones
Especificaciones
CD105 (Endoglin)
Monoclonal
PE
100 Tests
Human, Mouse
Hematopoietic Stem and Progenitor Cells, Mesenchymal Stem and Progenitor Cells
Immunoprecipitation, Western Blot, Flow Cytometry
43A3
Mouse
Primary
IgG1 κ
Documentos
One moment while we fetch your results.
Certificados
Promociones
Promociones
El Programa Encompass de Fisher Scientific ofrece artículos que no forman parte de nuestra cartera de distribución. Estos productos no suelen tener imágenes ni descripciones detalladas. Sin embargo, nos comprometemos a mejorar su experiencia de compra. Utilice el siguiente formulario para hacernos llegar sus comentarios sobre el contenido de este producto.
Título del producto
CD105 (Endoglin) Mouse, anti-Human, PE, Clone: 43A3, STEMCELL Technologies