Learn More
This item is not returnable.
View return policy
Description
- Etiquetas sin adhesivo de seguridad
- Sin adhesivo de bloqueo/etiquetado
Specifications
10/Paq. | |
TAG-SERVICE D'ENTRETIEN-75X160MM | |
Poliéster | |
Lockout Tagout | |
160 mm | |
3 in. | |
-40°C to +100°C | |
2.95 in. | |
7 mm |
Etiqueta de condenación | |
Blue on White | |
SERVICIO DE MANTENIMIENTO | |
6.3 in. | |
75 mm | |
0.249 mm | |
-40°F to +212°F | |
160 mm | |
Condenación |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Brady™ Etiquetas de precaución > Mensaje: Servicio de mantenimiento; Dimensiones (L x P): 75mm x 160mm
Spot an opportunity for improvement?Share a Content Correction