Learn More
Thermo Scientific™ CaptureSelect™ C-tagXL Affinity Matrix
Thermo Scientific™ CaptureSelect™ C-tagXL Affinity Matrix combines a unique selectivity for a small 4-amino acid peptide tag (E-P-E-A, glutamic acid-proline-glutamic acid-alanine) with the benefits of a robust and high quality affinity matrix.
Marca: Thermo Scientific™ 2943072010
Descripción
CaptureSelect C-tagXL Affinity Matrix combines a unique selectivity for a small 4-amino acid peptide tag (E-P-E-A, glutamic acid-proline-glutamic acid-alanine) with the benefits of a robust and high quality affinity matrix. It purifies C-terminal tagged proteins with high affinity and selectivity, even in the presence of urea and guanidine HCl, from complex mixtures like cell culture harvests and periplasmatic fractions in a one-step process. Mild elution conditions at neutral pH can be applied using magnesium chloride or propylene glycol, which ensures high activity recoveries of pH-sensitive target proteins. C-tagXL Affinity Matrix recognizes the E-P-E-A tag sequence when fused either directly to the C-terminus of a protein or through a linker between the C-terminus and the E-P-E-A tag.
Features of C-tagXL Affinity Matrix include:
- Mild elution, making it suitable for pH-sensitive proteins
- Binding of the tagged proteins under denaturing conditions (like dissolved inclusion bodies)
- Excellent scalability
- Non-animal-derived
CaptureSelect C-tagXL Affinity Matrix purifies C-terminal tagged E-P-E-A proteins directly from complex source materials in a single step with high purity and yield.
Free of animal components
CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).
Main characteristics:
- Matrix: agarose-based, epoxide-activated
- Average particle size: 65 ± 10 μm
- Ligand: CaptureSelect C-tagXL affinity ligand
- Ligand coupling method: epoxide coupling of the ligand
- Binding capacity: 400 nmol/mL resin depending on flow rate, column height, and contact time
- Elution conditions: Acidic: 20 mM citric acid or acetic acid, pH 3–4, or 0.1 M glycine pH 3.0; Neutral: 20 mM Tris, 2.0 M MgCl2 pH 7; 20 mM Tris, 1.0 M NaCl, 50% (v/v) propylene glycol; 20 mM Tris 2 mM S-E-P-E-A.
- Flow characteristics: 50–200 cm/h (up to 2 bar)
- Formulation buffer: 20%(v/v) ethanol
Especificaciones
400 nmol/mL | |
Agarose-based, epoxide-activated | |
Temperatura ambiente | |
Suspensión líquida | |
10 mL | |
C-tagXL affinity ligand |
50 to 200 cm/hr. (Up to 2 bar) | |
Store at 2°C to 8°C. | |
65 ±10 μm | |
CaptureSelect™ | |
Affinity Matrix | |
Anti-EPEA tag |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Para uso exclusivo en investigación. No apto para uso en procedimientos diagnósticos.