Learn More
ACROBiosystems Biotinylated Human BAFFR / TNFRSF13C Protein, His,Avitag™
MALS verified
Marca:  ACROBiosystems BAR-H82E3-200ug
Detalles adicionales : Peso : 0.00010kg
 Ver más versiones de este producto
Código de producto. 17186081
  • 1733.51€ / 200 microgramos
Fecha estimada de envío: 24-10-2022
para ver el stock
Descripción
Descripción
BAFF receptor (B-cell activating factor receptor, BAFF-R), also known as tumor necrosis factor receptor superfamily member 13C (TNFRSF13C), is a membrane protein of the TNF receptor superfamily which recognizes BAFF. B-cell activating factor (BAFF) enhances B-cell survival in vitro and is a regulator of the peripheral B-cell population. Overexpression of BAFF in mice results in mature B-cell hyperplasia and symptoms of systemic lupus erythematosus (SLE). Also, some SLE patients have increased levels of BAFF in serum. Therefore, it has been proposed that abnormally high levels of BAFF may contribute to the pathogenesis of autoimmune diseases by enhancing the survival of autoreactive B cells.
This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 10.1 kDa. The protein migrates as 15-22 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Especificaciones
Especificaciones
NP_443177.1
10.1 kDa
200 µg
Ser 7 - Ala 71
BAFFR,TNFRSF13C,BROMIX,CD268,CVID4,prolixin,BAFF-R
Precision Avi
PBS, pH7.4
HEK293
Human
Powder
>90% as determined by SDS-PAGE
Biotinylated Human BAFFR, His,Avitag (BAR-H82E3) is expressed from human 293 cells (HEK293). It contains AA Ser 7 - Ala 71 (Accession # Q96RJ3-1).
-20°C
BAFFR
His Tag and Avi Tag
Less than 1.0 EU / µg by the LAL method.
MABSol™ Biotin Labeled Proteins