missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Biotinylated Human BAFFR / TNFRSF13C Protein, His,Avitag™

MALS verified

419.88€ - 1847.47€

Especificaciones

Número de acceso NP_443177.1
Peso molecular 10.1 kDa
Fuente Biotinylated Human BAFFR, His,Avitag (BAR-H82E3) is expressed from human 293 cells (HEK293). It contains AA Ser 7 - Ala 71 (Accession # Q96RJ3-1).
Inmunógeno Ser 7 - Ala 71
Requisitos de almacenamiento -20°C
Ver más especificaciones

Productos 2
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
17186081
Ver documentos Detalles de la promoción
ACROBiosystems
BAR-H82E3-200UG
200 μg
1847.47€
200 microgramos
Ver productos alternativos
Ahorre hasta 
»
Fecha estimada de envío: 11-04-2024
para ver el stock
Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.
17196081
Ver documentos Detalles de la promoción
ACROBiosystems
BAR-H82E3-25UG
25 μg
419.88€
25 microgramos
Ver productos alternativos
Ahorre hasta 
»
Fecha estimada de envío: 11-04-2024
para ver el stock
Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.
Descripción

Descripción

BAFF receptor (B-cell activating factor receptor, BAFF-R), also known as tumor necrosis factor receptor superfamily member 13C (TNFRSF13C), is a membrane protein of the TNF receptor superfamily which recognizes BAFF. B-cell activating factor (BAFF) enhances B-cell survival in vitro and is a regulator of the peripheral B-cell population. Overexpression of BAFF in mice results in mature B-cell hyperplasia and symptoms of systemic lupus erythematosus (SLE). Also, some SLE patients have increased levels of BAFF in serum. Therefore, it has been proposed that abnormally high levels of BAFF may contribute to the pathogenesis of autoimmune diseases by enhancing the survival of autoreactive B cells.

This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 10.1 kDa. The protein migrates as 15-22 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Especificaciones

Especificaciones

NP_443177.1
Biotinylated Human BAFFR, His,Avitag (BAR-H82E3) is expressed from human 293 cells (HEK293). It contains AA Ser 7 - Ala 71 (Accession # Q96RJ3-1).
-20°C
BAFFR
Human
PBS, pH7.4
HEK293
MABSol™ Biotin Labeled Proteins
10.1 kDa
Ser 7 - Ala 71
BAFFR,TNFRSF13C,BROMIX,CD268,CVID4,prolixin,BAFF-R
Precision Avi
His Tag and Avi Tag
Less than 1.0 EU / μg by the LAL method.
Powder
>90% as determined by SDS-PAGE
Videos
SDS
Documentos

Documentos

Product Certifications