missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ BGN (Human) Recombinant Protein
Human BGN full-length ORF ( AAH02416.1, 1 a.a. - 368 a.a.) recombinant protein with GST-tag at N-terminal.
Marca: Abnova™ H00000633-P01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
- Sequence: MWPLWRLVSLLALSQALPFEQRGFWDFTLDDGPFMMNDEEASGADTSGVLDPDSVTPTYSAMCPFGCHCHLRVVQCSDLGLKSVPKEISPDTTLLDLQNNDISELRKDDFKGLQHLYALVLVNNKISKIHEKAFSPLRKLQKLYISKNHLVEIPPNLPSSLVELRIHDNRIRKVPKGVFSGLRNMNCIEMGGNPLENSGFEPGAFDGLKLNYLRISEAKLTGIPKDLPETLNELHLDHNKIQAIELEDLLRYSKLYRLGLGHNQIRMIENGSLSFLPTLRELHLDNNKLARVPSGLPDLKLLQVVYLHSNNITKVGVNDFCPMGFGVKRAYYNGISLFNNPVPYWEVQPATFRCVTDRLAIQFGNYKK
Especificaciones
AAH02416.1 | |
biglycan | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BGN | |
GST |
633 | |
Wheat germ expression system | |
25 μg | |
DSPG1, PG-S1, PGI, SLRR1A | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |