Learn More
Microbiologics™ Bacillus subtilis subsp. spizizenii ATCC™ 6633™†
For culture purposes and QC applications
Especificaciones
ATCC® | 6633 |
---|---|
Nivel de bioseguridad | 1 |
Certificaciones/conformidad | CE, online certificate of analysis |
Contenido y almacenamiento | De 2 °C a 8 °C |
Formulario | Lyophilized Pellet |
Descripción
KWIK-STIK™
- Clever design and accurate, repeatable results
- Each device features a single strain of Bacillus subtilis subsp. spizizenii in a lyophilized pellet, a reservoir of hydrating fluid and inoculating swab
- For added convenience, a peel-off identification label is included for easy documentation
- Available in packs of 2, 6 or 10
- The most economical reference stock culture option; lyophilized pellet containing a single strain of microorganism
- Easy to use; simply re-hydrate the pellet and inoculate
- Available in a re-sealable vial containing 6 or 10 pellets
- ATCC™ 6633™†
Especificaciones
6633 | |
CE, online certificate of analysis | |
Lyophilized Pellet | |
Bacillus subtilis |
1 | |
De 2 °C a 8 °C | |
Culture purpose and QC application |
Seguridad y manipulación
missing translation for 'shelfLife' : 6 to 24 Months
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. MicroBioLogics Inc. is licensed to use these trademarks and to sell products derived from ATCC cultures.
El Programa Encompass de Fisher Scientific ofrece artículos que no forman parte de nuestra cartera de distribución. Estos productos no suelen tener imágenes ni descripciones detalladas. Sin embargo, nos comprometemos a mejorar su experiencia de compra. Utilice el siguiente formulario para hacernos llegar sus comentarios sobre el contenido de este producto.