Learn More
B23/NPM1 Mouse anti-Human, CoraLite 594, Clone: 4F12A3, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech CL594-60096-100UL
Descripción
The B23/NPM1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human B23/NPM1. This antibody recognizes human, mouse, rat antigen. The B23/NPM1 antibody has been validated for the following applications: IF analysis. This antibody is coralite™594.
Especificaciones
B23/NPM1 | |
Monoclonal | |
500 μg/mL | |
Mouse | |
Primary | |
Human, Rat, Mouse |
Immunofluorescence | |
4F12A3 | |
CoraLite 594 | |
100 μL | |
4869 | |
IgG1 |