Learn More
Invitrogen™ Human AXL, His Tag Recombinant Protein
The purified recombinant protein can be used in a number of different applications, but it has been specifically validated for use with both the LanthaScreen™ & Z'-LYTE™ assay technologies to identify novel kinase inhibitors. AXL is also available in our SelectScreen™ Kinase Profiling Service.
Marca: Invitrogen™ A31516
Detalles adicionales : Peso : 0.01000kg
Descripción
AXL is a chronic myelogenous leukemia-associated oncogene that is also associated with colon cancer and melanoma. The AXL protein is a receptor tyrosine kinase that transduces signals from the extracellular matrix into the cytoplasm by binding growth factors like vitamin K-dependent protein growth-arrest-specific gene 6, and is involved in the stimulation of cell proliferation.
- Validated for use with the LanthaScreen & Z'-LYTE assay technology
- Available in multiple pack sizes for convenient use
- Also available in our SelectSceen Kinase Profiling Service
Especificaciones
NP_068713.2 | |
50mM sod¡um phosphate, pH 7.0, 300mM NaCl, 150mM imidazole, 0.1mM PMSF, 0.2mM DTT , 25% glycerol | |
100μg | |
Truncated, Recombinant, Cytoplasmic, Active | |
AXL Kinase Family | |
Human | |
His Tag |
Kinase Assay | |
48.3kDa | |
Store at -80°C. Upon first use, aliquot and store at -80°C to avoid multiple freeze/thaws. | |
UFO/JTK11 | |
473 - end | |
Recombinant | |
Insect |