Learn More
CPC (Colder) AseptiQuik™ STC II Genderless Quick-Disconnect Fittings, AspetiQuik™ S to Sanitary Adapter, Polycarbonate
Low-flow genderless sterile connector and Steam-Thru™ II connector in a single unit simplify process integration while maximizing flexibility
Marca:  CPC (Colder) 50112-04
 Ver más versiones de este producto
Código de producto. 30053916
  • 443.00€ / cada uno
Solo quedan null
Fecha estimada de envoi: 22-10-2025
Descripción
Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system. Free technical application support available to determine which connector type is needed for your application. Manufactured in an ISO Class 7 certified cleanroom to minimize contamination by pollutants. Simple flip, click, and pull design minimizes operator errors. Create quick and easy sterile connections, even in nonsterile environments. Robust construction provides reliable and repeatable fluid transfer. Specifically designed for small-volume fluid transfer applications. Innovative three-port steam design. Sterilize AseptikQuik STC II fitting on stainless steel equipment via SIP. 0.375 in. nominal flow path size
Especificaciones
Especificaciones
ADCF;FDA;Non-DEHP;Non-Phthalate;RoHS;USP
1°C
Quick Disconnect x Sanitary x Sanitary
Straight
Platinum-cured Silicone
1-1/2 in.
1 Ea.
130°C
3/4 in. Sanitary Clamp Size
White with blue pull tab and amber tint
Certificados
Promociones
Promociones
Título del producto
CPC (Colder) AseptiQuik™ STC II Genderless Quick-Disconnect Fittings, AspetiQuik™ S to Sanitary Adapter, Polycarbonate > 1-1/2 in.