missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ApoA1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA123059
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
This antibody does not cross-react with human Apolipoprotein A1. Predicted molecular weight of 31 kDa.
Apolipoprotein A-1 (apo A-1) in human plasma is a major protein component of high density lipoprotein (HDL). The molecular mass is 24 kDa. Apo A-1 is a cofactor for enzymes involved in the normal turnover of cholesterol and is produced in the liver. Plasma concentration is 270-380 mg/100 mL.
Especificaciones
ApoA1 | |
Polyclonal | |
Unconjugated | |
APOA1 | |
1-Sep; 2-Sep; Alp-1; Apo a; APO A1; apo A-1; APO I; apo(a); APOA1; Apoa-1; ApoAI; Apo-AI; apoA-I; Apolipoprotein; Apolipoprotein A1; apolipoprotein A-1; Apolipoprotein AI; apolipoprotein A-I; apolipoprotein A-I precursor; Apolipoprotein A-I(1-242); Brp-14; Ltw-1; Lvtw-1; MGC102525; MGC117399; MGC124929; preproapolipoprotein A-I; prepro-apolipoprotein-AI; ProapoA-I; Proapolipoprotein A-I; Sep-1; Sep2; Sep-2; Truncated apolipoprotein A-I; Udpgt; Ugt1; Ugt1a7 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
11806, 25081 | |
4° C, do not freeze | |
Liquid |
ELISA, Immunodiffusion, Western Blot | |
1 mg/mL | |
TBS with 1mM EDTA and 0.02% sodium azide; pH 8 | |
P04639, Q00623 | |
APOA1 | |
Purified mouse apolipoprotein AI from pooled mouse plasma high density lipoprotein. | |
100 μg | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
ApoA1 Polyclonal Antibody, Invitrogen™ > 100 μg; Unconjugated
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido