Learn More
Stat1 (N-Terminus) Mouse, Alexa Fluor 647, Clone: 1/STAT1, BD
Mouse Monoclonal Antibody
Marca: BD Biosciences 558560
273.19 EUR hasta el 2025-12-31
Use el código promocional "25339" para aplicar la oferta.
Descripción
Stat (Signal transducer and activators of transcription) proteins are critical mediators of the biologic activity of cytokines, including interleukins, interferons, erythropoietin, and growth factors. Ligand-receptor interaction leads to activation of constitutively associated JAK family kinases and subsequent recruitment/activation of Stat proteins by tyrosine phosphorylation. Active Stat proteins then move to the nucleus to promote transcription of cytokine-inducible genes. Seven Stat proteins have been cloned, each of which is differentially expressed and/or activated in a cytokine-specific and cell type-specific manner. Stat1 and Stat2 are components of the ISGF3 (Interferon-Stimulated Gene Factor 3) complex, which is the primary transcription activator induced by the binding of the interferon to a specific cell-surface receptor. Stat1 has two alternatively spliced isoforms, 91kDa Stat1α and 84kDa Stat1β Stat1α has 38 additional C-terminal amino acids. In response to the binding of IFNα, IFNγ, EGF, PDGF, or CSF-1 to their respective receptors, the Stat1 subunits become tyrosine-phosphorylated at Y701, and the complex is translocated to the nucleus. This results in the formation of an active complex that includes the DNA-binding p48 subunit. This complex is responsible for modulating the transcription of the interferon-stimulated genes (ISGs). Thus, phosphorylation of Y701 in Stat1 occurs in response to growth factors and cytokines, and is essential for normal transcriptional activity of the ISGF3 complex. The 1/Stat1 monoclonal antibody recognizes the N-terminus of human Stat1 (both isoforms), regardless of phosphorylation status.
Intracellular Staining

Especificaciones
Stat1 (N-Terminus) | |
Monoclonal | |
Alexa Fluor 647 | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
1/STAT1 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Human Stat1 aa. 1-194 | |
50 Tests | |
Cell Biology | |
Canine, Chicken, Frog, Human, Mouse, Rat | |
IgG1 |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
For Research Use Only.