Learn More
Ronin/THAP11 Mouse, Unlabeled, Clone: P56-507, BD
Mouse Monoclonal Antibody
Marca: BD Biosciences 562548
Descripción
Thanatos-associated proteins (THAPs) are characterized by the THAP domain, which is a conserved Drosophila P element transposase DNA-binding domain. THAPs are DNA-binding factors involved in cell proliferation, apoptosis, cell cycle and transcriptional regulation. THAP11 is an essential factor in embryogenesis and ES cell pluripotency and binds directly to host cell factor 1 (HCF-1), a key regulator of transcriptional control. THAP11 has been designated as Ronin, named for a masterless Japanese samurai because of its lack of any apparent relationship to Nanog, Oct4 and Sox2–the master regulators of pluripotency. Ronin/THAP11 binds to the c-Myc promoter and down-regulates c-Myc expression. Recently, it was reported that Bcr-Abl inhibited the expression of THAP11 in Chronic myelogenous leukemia (CML) cells and promoted CML cell proliferation by the aberrant induction of c-Myc expression.
Host Species: Mouse
Clone: P56-507
Isotype: IgG2b, κ
Species Reactivity: Human
Immunogen: Human Ronin/THAP11 Recombinant Protein
Western Blotting

Especificaciones
Ronin/THAP11 | |
Monoclonal | |
0.5mg/mL | |
Ronin, THAP11 | |
Mouse | |
Affinity Purified | |
RUO | |
Human, Mouse | |
IgG2b κ |
Western Blot | |
P56-507 | |
Unconjugated | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Human Ronin/THAP11 Recombinant Protein | |
0.1 mg | |
Primary | |
Store undiluted at 4°C |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.