Learn More
Rae-1 Pan Specific Rat anti-Mouse, Clone: 186107, R&D Systems™

Rat Monoclonal Antibody has been used in 18 publications
Brand: R&D Systems MAB17582-100

Description
Rae-1 Monoclonal specifically detects Rae-1 in Mouse samples. It is validated for Flow Cytometry, CyTOF-ready.Specifications
Rae-1 | |
Monoclonal | |
LYOPH | |
Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
Q9JI58 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
8480 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Flow Cytometry, CyTOF | |
186107 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Raet1a | |
Mouse myeloma cell line NS0-derived recombinant mouse Rae-1 delta Leu29-Ser227 Accession # Q9JI58 | |
100 μg | |
Primary | |
Detects mouse Rae-1. It recognizes Rae-1 alpha , beta , delta , gamma and epsilon . | |
Mouse | |
Supernatant |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
For Research Use Only