missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDEF Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen 363600
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
PDEF (prostate-derived Ets factor) is a 37.5-kDa protein that acts as an androgen-independent transcriptional activator for the PSA (prostate specific antigen) promoter. It also interacts directly with the DNA binding domain of androgen receptor and enhances androgen-mediated activation of the PSA promoter. PDEF was shown to have transcript levels 192-fold higher in the peripheral blood of some breast cancer patients in comparison with normal individuals. PDEF mRNA is also frequently over-expressed in human breast tumors as well as in human ovarian tumors. In ovarian tumors, PDEF expression seems to down-modulate maligt potential, and thus provides a rationale to screen for drugs that induce PDEF expression in epithelial ovarian tumors.
Especificaciones
PDEF | |
Polyclonal | |
Unconjugated | |
O95238 | |
SPDEF | |
Synthetic peptide derived from the internal region of human PDEF (prostate-derived Ets factor). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
ELISA, Western Blot | |
0.25 mg/mL | |
SPDEF | |
SAM pointed domain-containing Ets transcription factor, Prostate epithelium-specific Ets transcription factor, Prostate-specific Ets, Prostate-derived Ets factor, SPDEF, PDEF, PSE | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
25803 | |
-20°C | |
Liquid |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
PDEF Rabbit anti-Human, Polyclonal, Invitrogen™ > 100 μg; Unconjugated
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido