Learn More
Lambda, Ig Light Chain Mouse anti-Human, Unlabeled, Clone: 1-155-2, BD
Mouse Monoclonal Antibody
Marca: BD Biosciences 345130
Descripción
Anti-Lambda is specific for lambda light chains of human immunoglobulins. Immunoglobulins bearing lambda light chains are present on approximately 40% of normal B lymphocytes and on Igλ+ neoplastic cells. In serum, Anti-Lambda reacts with immunoglobulins bearing lambda light chains as well as free lambda light chains.
Host Species: Mouse
Clone: 1-55-2
Isotype: IgG1, λ
Cross Reactivity: Human
Flow Cytometry

Especificaciones
Lambda, Ig Light Chain | |
Monoclonal | |
50μg/mL | |
Phosphate buffered saline with gelatin and 0.1% sodium azide. | |
Human IgA | |
100 μg | |
Immunology | |
Human | |
IgG1 λ |
Flow Cytometry | |
1-155-2 | |
Unconjugated | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed. |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
For Research Use Only.