Learn More
anti-IL-21, PE, Clone: 149204, R&D Systems™
Rat Monoclonal Antibody has been used in 2 publications
Marca: R&D Systems IC594P-025

Descripción
IL-21 Monoclonal specifically detects IL-21 in Mouse samples. It is validated for Intracellular Staining by Flow Cytometry.Especificaciones
IL-21 | |
Monoclonal | |
R-PE | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
CVID11, IL21, IL-21Za11interleukin-21, interleukin 21, interleukin-21 isoform, Za11 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
59067 | |
Mouse | |
Purified |
Flow Cytometry | |
149204 | |
Intracellular Staining by Flow Cytometry 10 uL/10^6 cells | |
Q9ES17 | |
IL21 | |
E. coli-derived recombinant mouse IL-21 Pro25-Ser146 Accession # Q9ES17 | |
25 Tests | |
Primary | |
Detects mouse IL-21 in Western blots. Does not cross-react with recombinant human IL-21, recombinant mouse (rm) IL-15, or rmIL-12. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG2b |
El Programa Encompass de Fisher Scientific ofrece artículos que no forman parte de nuestra cartera de distribución. Estos productos no suelen tener imágenes ni descripciones detalladas. Sin embargo, nos comprometemos a mejorar su experiencia de compra. Utilice el siguiente formulario para hacernos llegar sus comentarios sobre el contenido de este producto.