Learn More
HLA-DR Mouse anti-Human, NA/LE, Unlabeled, Clone: L243 (G46-6), BD
Mouse Monoclonal Antibody
Marca: BD Biosciences 555809
375.77 EUR hasta el 2025-12-31
Use el código promocional "25339" para aplicar la oferta.
Descripción
Reacts with HLA-DR, a human class II antigen II of the major histocompatibility complex (MHC). HLA-DR is a transmembrane glycoprotein composed of an α chain (36kDa) and a β subunit (27kDa) expressed primarily on antigen presenting cells: B cells, monocytes, macrophages, and thymic epithelial cells. HLA-DR is also expressed on activated T cells. This molecule plays a major role in cellular interaction during antigen presentation. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Host Species: Mouse
Clone: G46-6
Isotype: IgG2a κ
Species Reactivity: Human
Flow Cytometry

Especificaciones
HLA-DR | |
Monoclonal | |
1.0mg/mL | |
No azide/low endotoxin: Aqueous buffered solution containing no preservative, 0.2μm sterile filtered. Endotoxin level is ≤0.01 EU/μg (≤0.001 ng/μg) of protein as determined by the LAL assay. | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C |
Flow Cytometry | |
L243 (G46-6) | |
Unconjugated | |
Mouse | |
0.5 mg | |
Immunology | |
Baboon, Canine, Cynomolgus, Human, Rhesus Monkey | |
IgG2a κ |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
For Research Use Only.