Learn More
Histone H3 (Citrulline Arg17, Citrulline Arg2, Citrulline Arg8) Rabbit anti-Human, Mouse, Rat, Bacteria, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 6 publications
Marca: Novus Biologicals NB100-57135

Descripción
Histone H3 Polyclonal specifically detects Histone H3 in Human, Mouse, Rat, Bacteria samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Especificaciones
Histone H3 (Citrulline Arg17, Citrulline Arg2, Citrulline Arg8) | |
Polyclonal | |
Western Blot 1:5 000 - 1:50000, Chromatin Immunoprecipitation 1:10-1:500, Flow Cytometry, ELISA, Immunohistochemistry 1:20000 - 1:50000, Immunocytochemistry/Immunofluorescence 1:2 000 - 1:20 000, Immunohistochemistry-Paraffin 1:20000 - 1:50000, Immunohistochemistry-Frozen | |
H3, H3 histone, family 3A, H3.3A, H3.3B, H3F2, H3F3, H3F3B, H3FM, HIST2H3C, histone H3.3, MGC87782, MGC87783 | |
Rabbit | |
15 kDa | |
0.2 mL | |
Primary | |
This Histone H3 antibody reacts specifically with human Histone H3 Citrulline at either position 2 and or 8 and or 17. Not yet tested in other species, but the should react with a wide range of species due to sequence homology. | |
Human, Mouse, Rat, Bacteria | |
IgG |
Western Blot, ChIP assay, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
Unconjugated | |
Antiserum with No Preservative | |
H3C14 | |
This Histone H3 antibody was raised against synthetic peptide derived from N terminal region of Human Histone H3, containing Citrulline at position 2;8;17 | |
Unpurified | |
RUO | |
126961 | |
Please note the larger size(s) come in lyophilized form and the sample size will be in liquid formate. Reconstitute with 0.2 ml distilled water. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
El Programa Encompass de Fisher Scientific ofrece artículos que no forman parte de nuestra cartera de distribución. Estos productos no suelen tener imágenes ni descripciones detalladas. Sin embargo, nos comprometemos a mejorar su experiencia de compra. Utilice el siguiente formulario para hacernos llegar sus comentarios sobre el contenido de este producto.
For Research Use Only