Learn More
Alpha-fetoprotein Mouse anti-Human, PE, Clone: C3/AFP, BD
Mouse Monoclonal Antibody
Marca: BD Biosciences 563002
Descripción
Alpha-fetoprotein (AFP) is a member of the albuminoid gene family which includes albumin, alpha-albumin, and vitamin D protein. Alpha-fetoprotein is an abundant plasma protein synthesized from fetal yolk sac, liver, and gastrointestinal tract during development. Similar to albumin, AFP binds and transports multiple ligands such as nickel, copper, billirubin, and fatty acids. AFP levels are low to absent in healthy adult tissues whereas elevated AFP levels in the adult can be indicative of malignancies. In particular, AFP is expressed in hepatocellular carcinoma, germ cell tumors, and metatstatic cancers of the liver. Maternal serum AFP levels can be monitored to screen for fetal abnormalities. AFP is used as a hepatic progenitor marker in hepatic differentiations from pluripotent stem cells.

Especificaciones
Alpha-fetoprotein | |
Monoclonal | |
PE | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Human Alpha-fetoprotein | |
50 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
C3/AFP | |
Alpha-1-fetoprotein, Alpha-fetoglobulin, AFP | |
Mouse | |
Affinity Purified | |
RUO | |
Human | |
IgG2a |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.