missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ENDOG Monoclonal Antibody (7F2G10), Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen MA515759
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
A suggested positive control is HepG2 cell lysate.
The protein encoded by this gene is a nuclear encoded endonuclease that is localized in the mitochondrion. The encoded protein is widely distributed among animals and cleaves DNA at GC tracts. This protein is capable of generating the RNA primers required by DNA polymerase gamma to initiate replication of mitochondrial DNA.
Especificaciones
ENDOG | |
Monoclonal | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q14249 | |
ENDOG | |
Recombinant protein corresponding to amino acids 51 - 140 of human EndoG. | |
100 μg | |
Primary | |
Human, Rat | |
Antibody | |
IgG1 |
Western Blot | |
7F2G10 | |
Unconjugated | |
ENDOG | |
endo G; Endog; endonuclease G; endonuclease G, mitochondrial; mitochondrial endonuclease G | |
Mouse | |
Protein A | |
RUO | |
2021, 362100 | |
-20°C | |
Liquid |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
ENDOG Monoclonal Antibody (7F2G10), Invitrogen™ > 100 μg; Unconjugated
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido