Learn More
Cytochrome c Mouse, Unlabeled, Clone: 7H8.2C12, BD
Mouse Monoclonal Antibody
Marca: BD Biosciences 556433
218.12 EUR hasta el 2025-12-31
Use el código promocional "25339" para aplicar la oferta.
Descripción
A cytochrome is an electron-transporting protein that contains a heme prosthetic group. Cytochromes have been known to be essential components of the mitochondrial respiratory chain since 1925. The iron atom of the heme group in cytochromes alternates between a reduced ferrous (+2) state and an oxidized ferric (+3) state during electron transport in oxidative phosphorylation. Cytochromes are classified into four groups (a, b, c, and d) according to spectrochemical characteristics, and there are five cytochromes between Coenzyme QH2 and O2 in the electron transport chain. Cytochrome c is a water-soluble peripheral membrane protein of the mitochondria and transports electrons from the Coenzyme QH2 - Cytochrome c reductase complex to the Cytochrome c oxidase complex. Recently it has been suggested that Cytochrome c may be required for cells to undergo apoptosis. Mitochondria may be involved in apoptosis by releasing Cytochrome c to the cytosol where it functions in the activation of Caspase 3 (CPP32), an ICE family molecule that has been shown to trigger apoptosis when activated. In the cytoplasm Cytochrome C might exist in polymeric forms, appearing as a 58-60kDa band by western blot analysis, rather than the monomeric form. Monomeric Cytochrome c migrates at a reduced molecular weight of 15kDa. Clone 7H8.2C12 recognizes the denatured form of pigeon, horse, rat, mouse and human Cytochrome c. It does not recognize the native form of Cytochrome c. Clone 7H8.2C12 recognizes an epitope within amino acids 93-104 (inclusive) of pigeon Cytochrome C, based on competitive ELISA results. Synthetic peptides corresponding to amino acids 1-80, 81-104 and 66-104 of pigeon Cytochrome c were used as immunogens. Clone 7H8.2C12 is used for western blot detection of Cytochrome c. Clone 6H2.B4, Mfr. No. 556432, is used for immunoprecipitation. Cell lines P388D1 mouse lymphoma (ATCC CCL-46), HeLa human carcinoma (ATCC CCL-2), Jurkat T leukemia (ATCC TIB-152), and NIH/3T3 mouse fibroblast cells (ATCC CRL-1658) may be used as positive control cell lysates. This antibody is routinely tested by western blot analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Host Species: Mouse
Clone: 7H8.2C12
Isotype: IgG2b, κ
Species Reactivity: Human
Immunogen: Synthetic Peptides of Pigeon Cytochrome C
Western Blotting

Especificaciones
Cytochrome c | |
Monoclonal | |
0.5mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Synthetic Peptides of Pigeon Cytochrome C | |
0.1 mg | |
Cell Biology | |
Human, Mouse, Pigeon, Rat | |
IgG2b κ |
Western Blot | |
7H8.2C12 | |
Unconjugated | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
For Research Use Only.