missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-CD8 alpha FITC, Clone: 53-6.7, Invitrogen™
CD8 alpha Monoclonal antibody A18649 from Invitrogen specifically detects CD8 alpha in Mouse samples. It is validated for Flow Cytometry.
Marca: Invitrogen A18649
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
CD8 alpha Monoclonal antibody A18649 from Invitrogen specifically detects CD8 alpha in Mouse samples. It is validated for Flow Cytometry.
The CD8 antigen is a cell surface glycoprotein found on most cytotoxic T lymphocytes that mediates efficient cell-cell interactions within the immune system. The CD8 antigen acts as a corepressor with the T-cell receptor on the T lymphocyte to recognize antigens displayed by an antigen presenting cell (APC) in the context of class I MHC molecules. The coreceptor functions as either a homodimer composed of two alpha chains, or as a heterodimer composed of one alpha and one beta chain. Both alpha and beta chains share significant homology to immunoglobulin variable light chains. This gene encodes the CD8 alpha chain isoforms. Multiple transcript variants encoding different isoforms have been found for this gene.Host Species: Rat
Cross Reactivity: Mouse
Isotype: IgG2a, κ
Clone: 53-6.7
Immunogen: Mouse CD8a.
Formulation: 25μg Purified Antibody (0.2mg/ml) in PBS, with 0.1% gelatin and 0.09% sodium azide; pH7.2
Synonyms: CD8, Leu-2, MAL, p32, CD8a
Uniprot Accession: P01731
Entrez GeneID: 12525
Gene Symbol: Cd8a
This product detects CD8 alpha in Mouse samples. This product has been successfully used in Flow Cytometry procedures. The immunogen is Mouse CD8a.

Especificaciones
CD8 alpha | |
Monoclonal | |
FITC | |
P01731 | |
Cd8a | |
Mouse CD8a. | |
Primary | |
Mouse | |
Purified |
Flow Cytometry | |
53-6.7 | |
25μg Purified Antibody (0.2mg/ml) in PBS, with 0.1% gelatin and 0.09% sodium azide; pH7.2 | |
CD8, Leu-2, MAL, p32, CD8a | |
Rat | |
25 μg | |
12525 | |
Store at 4° C | |
IgG2a κ |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
anti-CD8 alpha FITC, Clone: 53-6.7, Invitrogen™ > 25μg; FITC
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido