Learn More
CD66c Mouse anti-Human, PE, Clone: B6.2/CD66C, BD
Mouse Monoclonal Antibody
Marca: BD Biosciences 551478
Descripción
Reacts with two glycosylphosphatidylinositol-anchored glycoprotein present on granulocytes. Antibody B6.2 was studied as recognizing CD66c in the VI Human Leukocyte Differentiation Antigen workshop. CD66 antigens also known as the carcinoembryonic antigen (CEA) family of molecules, are closely related to the immunoglobulin super family of glycoproteins. Studies on CD66 molecules suggest a potential adhesion function in vivo. These molecules exhibit both homophilic and heterophilic adhesion. CEA family members may be involved intransmembrane signalling and activation of neutrophils. This clone has been found to be N-terminal domain reactive, reacted preferentially with the native protein and were conformationally dependent.

Especificaciones
CD66c | |
Monoclonal | |
PE | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
B6.2/CD66C | |
CEA, carcinoembryonic antigen | |
Mouse | |
100 Tests | |
Immunology | |
Human | |
IgG1 κ |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
For Research Use Only.