missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ANGPTL7 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
121.38€ - 325.38€
Especificaciones
Antígeno | ANGPTL7 |
---|---|
Concentración | 0.17 mg/mL |
Aplicaciones | Western Blot |
Clasificación | Polyclonal |
Conjugado | Unconjugated |
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
16826763
|
Proteintech
10396-1-AP-20UL |
20 μL |
Este artículo actualmente no está disponible o ha sido discontinuado. Ver la página del producto para posibles alternativas |
||||||
16816763
|
Proteintech
10396-1-AP-150UL |
150 μL |
Este artículo actualmente no está disponible o ha sido discontinuado. Ver la página del producto para posibles alternativas |
||||||
Descripción
Angptl7 (angiopoietin-like 7), alternately known as cornea-derived transcript 6 protein (CDT6) or AngX, is a 346 amino acid secreted protein that is expressed specifically in the stromal layer of cornea. Angptl7 forms disulfide-linked homotetramers via coiled-coil interactions and contains one fibrinogen C-terminal domain. Overexpressed in patients with glaucoma, Angptl7 serves as a potential therapeutic target for glaucoma and has been found to increase collagen expression. The gene encoding Angptl7 maps to human chromosome 1, which spans 260 million base pairs, contains over 3,000 genes and comprises nearly 8% of the human genome.Especificaciones
ANGPTL7 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Mouse, Human | |
O43827, Q8R1Q3 | |
10218, 654812 | |
ANGPTL7 Fusion Protein Ag0596 | |
Primary | |
-20°C | |
ANGPTL7 |
0.17 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
angiopoietin like 7, Angiopoietin like factor, Angiopoietin like protein 7, Angiopoietin related protein 7, ANGPTL7, AngX, CDT6, RP4 647M16.2 | |
Angptl7 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |