missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ANGPTL7 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 10396-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
Angptl7 (angiopoietin-like 7), alternately known as cornea-derived transcript 6 protein (CDT6) or AngX, is a 346 amino acid secreted protein that is expressed specifically in the stromal layer of cornea. Angptl7 forms disulfide-linked homotetramers via coiled-coil interactions and contains one fibrinogen C-terminal domain. Overexpressed in patients with glaucoma, Angptl7 serves as a potential therapeutic target for glaucoma and has been found to increase collagen expression. The gene encoding Angptl7 maps to human chromosome 1, which spans 260 million base pairs, contains over 3,000 genes and comprises nearly 8% of the human genome.Especificaciones
ANGPTL7 | |
Polyclonal | |
Unconjugated | |
ANGPTL7 | |
angiopoietin like 7, Angiopoietin like factor, Angiopoietin like protein 7, Angiopoietin related protein 7, ANGPTL7, AngX, CDT6, RP4 647M16.2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10218, 654812 | |
-20°C | |
Liquid |
Western Blot | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O43827, Q8R1Q3 | |
Angptl7 | |
ANGPTL7 Fusion Protein Ag0596 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |