missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Adenosine A1 Receptor Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 20332-1-AP-150UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Adenosine receptors (ARs) are members of the 7-transmembrane domain G-protein-coupled receptor superfamily. Structural, biochemical and pharmacological analyses of the AR genes and protein has led to the discovery of four distinct AR subtypes (A1, A2a, A2b, A3). Activation of ARs mediates several receptor subtype-specific physiological processes that include cardiac rate, smooth muscle tone, platelet aggregation, inflammation, cell growth and death, and neurotransmission. The A1AR is a glycoprotein that can activate Gi and Go proteins in vitro. In intact cells, agonist occupation of the A1AR has been shown to cause pertussis toxin-sensitive inhibition of adenylyl cyclase activity and, in some systems, a stimulation of phospholipase C resulting in mobilization of intracellular calcium stores. Activation of potassium channels by A1AR has been intensively studied in relation to its dramatic effects on the cardiovascular system. A1AR protein is highly expressed in brain (especially cerebellum, hippocampus, thalamus, and cortex) and spinal cord and in part, modulates neurotransmitter release. In white adipocytes A1AR inhibits lipolysis and stimulates glucose uptake. Other tissues also express A1AR including kidney and testis.Especificaciones
Adenosine A1 Receptor | |
Polyclonal | |
Unconjugated | |
ADORA1 | |
adenosine A1 receptor, Adenosine receptor A1, ADORA1, RDC7 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
11539, 134 | |
-20°C | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
0.15 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P30542, Q60612 | |
ADORA1 | |
Adenosine A1 Receptor Fusion Protein Ag14092 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Adenosine A1 Receptor Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 150 μL; Unconjugated
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido