missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABC2 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Marca: Novus Biologicals NB100-74598CL1
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
ABC2 Polyclonal antibody specifically detects ABC2 in Human samples. It is validated for Immunohistochemistry,Immunoprecipitation,Immunohistochemistry (Paraffin),Western Blot (Negative)
Especificaciones
ABC2 | |
Polyclonal | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot (Negative) | |
CoraFluor 1 | |
ABC2, Amplified in breast cancer gene 2 protein, chromosome 17 open reading frame 71, FLJ10587, protein SMG8, Protein smg-8 homolog, SMG8 | |
The immunogen recognized by this antibody maps to a region between residue 250 and 300 of human amplified in breast cancer gene 2 protein using the numbering given in entry NP_060619.4 (GeneID 55181). | |
0.1 mL | |
Cancer | |
55181 | |
Store at 4°C in the dark. Do not freeze. | |
IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
ABC2 Antibody, CoraFluor™ 1, Novus Biologicals™ > 0.1 mL; CoraFluor 1
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido