missing translation for 'onlineSavingsMsg'
Learn More
Learn More
A33 Recombinant Rabbit Monoclonal Antibody (4H4), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Marca: Invitrogen MA549954
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
The glycoprotein encoded by this gene is a cell surface antigen that is expressed in greater than 95% of human colon cancers. The open reading frame encodes a 319-amino acid polypeptide having a putative secretory signal sequence and 3 potential glycosylation sites. The predicted mature protein has a 213-amino acid extracellular region, a single transmembrane domain, and a 62-amino acid intracellular tail. The sequence of the extracellular region contains 2 domains characteristic of the CD2 subgroup of the immunoglobulin (Ig) superfamily.
Especificaciones
A33 | |
Recombinant Monoclonal | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide, pH 7.4 | |
Q99795 | |
Gpa33 | |
A synthesized peptide derived from human GPA33 | |
100 μL | |
Primary | |
Human | |
Liquid |
ELISA, Immunohistochemistry (Paraffin) | |
4H4 | |
Unconjugated | |
Gpa33 | |
2010310L10Rik, 2210401D16Rik, A33, A33 antigen, BB116197, cell surface A33 antigen, Glycoprotein A33, glycoprotein A33 (transmembrane), GPA33, mA33, MGC129986, MGC129987, transmembrane glycoprotein A33 | |
Rabbit | |
Affinity chromatography | |
RUO | |
10223 | |
-20°C or -80°C if preferred | |
IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
A33 Recombinant Rabbit Monoclonal Antibody (4H4), Invitrogen™ > 100 μL; Unconjugated
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido