missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDT2 Antibody (JE58-01), Novus Biologicals™
Rabbit Monoclonal Antibody
Marca: Novus Biologicals NBP3-32176
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
CDT2 Monoclonal antibody specifically detects CDT2 in Human,Mouse samples. It is validated for Flow Cytometry,Western Blot
Especificaciones
CDT2 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:2000, Flow Cytometry 1:1000 | |
CDT2RA-regulated nuclear matrix-associated protein, CDW1, DCAF2DDB1 and CUL4 associated factor 2, DDB1- and CUL4-associated factor 2, denticleless homolog (Drosophila), L2DTLRA regulated nuclear matrix associated protein, Lethal(2) denticleless protein homolog, RAMPdenticleless protein homolog, Retinoic acid-regulated nuclear matrix-associated protein | |
Recombinant protein within Human CDT2 aa 1-200/ 730. (Uniprot: Q9NZJ0) | |
100 μL | |
Cell Cycle and Replication, Core ESC Like Genes, Stem Cell Markers | |
51514 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Flow Cytometry, Western Blot | |
JE58-01 | |
Unconjugated | |
1*TBS (pH7.4), 0.05% BSA and 40% Glycerol | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
CDT2 Antibody (JE58-01), Novus Biologicals™ > 100 μL; Unconjugated
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido