missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MHC Class II RT1B Antibody (OX-6) [Janelia Fluor™ 669], Novus Biologicals™
Mouse Monoclonal Antibody
Marca: Novus Biologicals (Bio-Techne) NB100-65541JF669
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
MHC Class II RT1B Monoclonal antibody specifically detects MHC Class II RT1B in Mouse,Rat samples. It is validated for Flow Cytometry,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunohistochemistry (Frozen),Immunohistochemistry Free-Floating,CyTOF,Immunocytochemistry/ Immunofluorescence
Especificaciones
MHC Class II RT1B | |
Monoclonal | |
Janelia Fluor 669 | |
DMA, DMB, DP beta 1, DPB 1, DPB1, DRB, HLA class II histocompatibility antigen DM beta chain, HLA DMB, HLA DPB1, HLADM, HLADP1B, Major histocompatibility complex class II, Major histocompatibility complex class II DP beta 1, MHC class II antigen DMB, MHC DPB1, RING6, RING7 | |
Rat thymocyte membrane glycoproteins | |
0.25 mL | |
Adaptive Immunity, Immunology | |
24738 | |
Store at 4°C in the dark. | |
IgG1 |
Flow Cytometry, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunohistochemistry (Free Floating), CyTOF, Immunocytochemistry/Immunofluorescence | |
OX-6 | |
50mM Sodium Borate | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Mouse, Rat | |
Purified |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
MHC Class II RT1B Antibody (OX-6) [Janelia Fluor™ 669], Novus Biologicals™ > 0.25 mL; Janelia Fluor 669
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido