missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ MR1 Antibody (MR1/7577) [Alexa Fluor™ 405], Novus Biologicals™
Mouse Monoclonal Antibody
Marca: Novus Biologicals™ NBP3-26943AF405
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
MR1 Monoclonal antibody specifically detects MR1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Especificaciones
MR1 | |
Monoclonal | |
Alexa Fluor 405 | |
Class I histocompatibility antigen-like protein, FLJ31593, HLALS, major histocompatibility complex class I-related gene protein, major histocompatibility complex, class I-like sequence, major histocompatibility complex, class I-related, MHC class I-like antigen MR-1, MHC class I-related gene protein, MHC class-I related-gene protein | |
Recombinant fragment (around aa50-250) of human MR1 protein (exact sequence is proprietary) | |
0.1 mL | |
Immunology | |
3140 | |
Store at 4°C in the dark. | |
IgG2 κ |
Immunohistochemistry (Paraffin) | |
MR1/7577 | |
50 mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Novus Biologicals™ MR1 Antibody (MR1/7577) [Alexa Fluor™ 405], Novus Biologicals™ > 0.1 mL, Alexa Fluor 405
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido