missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mesothelin Antibody (CL13956) [Biotin], Novus Biologicals™
Mouse Monoclonal Antibody
Marca: Novus Biologicals NBP3-24589B
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Mesothelin Monoclonal antibody specifically detects Mesothelin in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Especificaciones
Mesothelin | |
Monoclonal | |
Biotin | |
CAK1, CAK1 antigen, megakaryocyte potentiating factor, mesothelin, MPFSMRP, Pre-pro-megakaryocyte-potentiating factor, soluble MPF mesothelin related protein | |
This antibody was developed using a recombinant protein derived from Q13421, with the exact immunogen sequence remaining proprietary. | |
0.1 mL | |
Cancer | |
10232 | |
Store at 4⪚C in the dark. | |
IgG1 |
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
CL13956 | |
PBS | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Mesothelin Antibody (CL13956) [Biotin], Novus Biologicals™ > 0.1 mL; Biotin
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido