missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CS Citrate Synthase Antibody (CL2579) [Alexa Fluor™ 405], Novus Biologicals™
Mouse Monoclonal Antibody
Marca: Novus Biologicals NBP2-36775AF405
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
CS Citrate Synthase Monoclonal antibody specifically detects CS Citrate Synthase in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Especificaciones
CS Citrate Synthase | |
Monoclonal | |
Alexa Fluor 405 | |
citrate synthase, citrate synthase, mitochondrial, EC 2.3.3, EC 2.3.3.1 | |
This antibody was developed using a recombinant protein derived from O75390, with the exact immunogen sequence remaining proprietary. | |
0.1 mL | |
Cancer, Endocrinology, Signal Transduction | |
1431 | |
Store at 4⪚C in the dark. | |
IgG1 |
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
CL2579 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
CS Citrate Synthase Antibody (CL2579) [Alexa Fluor™ 405], Novus Biologicals™ > 0.1 mL; Alexa Fluor 405
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido