missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dopamine D1R/DRD1 Antibody (SG2-D1a), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Marca: Novus Biologicals NB110-60017JF646
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Dopamine D1R/DRD1 Monoclonal antibody specifically detects Dopamine D1R/DRD1 in Mouse, Rat, Bovine (Negative), Canine (Negative), Human (Negative), Porcine (Negative), Rabbit (Negative), Sheep (Negative) samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry Free-Floating
Especificaciones
Dopamine D1R/DRD1 | |
Monoclonal | |
Janelia Fluor 646 | |
D(1A) dopamine receptor, D1DR, DADR, Dopamine D1 receptor, dopamine receptor D1, DRD1A | |
Recombinant rat Dopamine Receptor D1 protein near the C-terminus. [Swiss-Prot# P18901] | |
0.1 mL | |
GPCR, Neuroscience, Neurotransmission | |
1812 | |
Store at 4°C in the dark. | |
IgG2b κ |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Free Floating) | |
SG2-D1a | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Mouse, Rat, Bovine (Negative), Canine (Negative), Human (Negative) | |
Purified |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Dopamine D1R/DRD1 Antibody (SG2-D1a), Janelia Fluor™ 646, Novus Biologicals™ > 0.1 mL; Janelia Fluor 646
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido