missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Complement C3 Antibody (NGM621), CoraFluor™ 1, Novus Biologicals™
Human Monoclonal Antibody
Marca: Novus Biologicals NBP3-28216CL1
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Complement C3 Monoclonal antibody specifically detects Complement C3 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Especificaciones
Complement C3 | |
Monoclonal | |
CoraFluor 1 | |
Acylation Stimulating Protein, acylation-stimulating protein cleavage product, AHUS5, ARMD9, ASP, C3, C3 and PZP-like alpha-2-macroglobulin domain-containing protein 1, C3a, C3a anaphylatoxin, C3adesArg, C3b, C3bc, C3-beta-c, complement C3, Complement C3 alpha chain, Complement C3 beta chain, Complement C3b alpha' chain, Complement C3c alpha' chain fragment 1, Complement C3c alpha' chain fragment 2, Complement C3d fragment, Complement C3dg fragment, Complement C3f fragment, Complement C3g fragment, complement component 3, complement component C3, complement component C3a, complement component C3b, CPAMD1, EC 3.4.21.43, epididymis secretory sperm binding protein Li 62p, HEL-S-62p, prepro-C3 | |
Complement C3 | |
0.1 mL | |
Angiogenesis, Apoptosis, Autoimmune Diseases, Immunology, Inflammation, Innate Immunity, Mesenchymal Stem Cell Markers, Neurodegeneration, Neuroscience, Signal Transduction | |
718 | |
Store at 4°C in the dark. Do not freeze. | |
IgG1 |
Flow Cytometry, ELISA, Functional Assay | |
NGM621 | |
PBS | |
Human | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Complement C3 Antibody (NGM621), CoraFluor™ 1, Novus Biologicals™ > 0.1 mL; CoraFluor 1
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido