missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Common gamma Chain/IL-2 R gamma Antibody (633162), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Marca: Novus Biologicals FAB2842Y
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Common gamma Chain/IL-2 R gamma Monoclonal antibody specifically detects Common gamma Chain/IL-2 R gamma in Human samples. It is validated for Flow Cytometry, Neutralization, CyTOF
Especificaciones
Common gamma Chain/IL-2 R gamma | |
Monoclonal | |
DyLight 680 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry, Neutralization, CyTOF | |
633162 | |
Flow Cytometry, Neutralization, CyTOF-ready | |
CD132, CD132 antigen, CIDX, combined immunodeficiency, X-linked, common cytokine receptor gamma chain, common gamma-chain, cytokine receptor common subunit gamma, gamma(c), IL-2 receptor subunit gamma, IL-2R subunit gamma, IL-2RG, IMD4, interleukin 2 receptor, gamma, Interleukin-2 receptor subunit gamma, p64, SCIDX, SCIDX1, severe combined immunodeficiency | |
S. frugiperda insect ovarian cell line Sf21-derived recombinant human Common gamma Chain/IL-2 gamma , Leu23-Asn254, Accession # P31785 | |
0.1 mL | |
Cancer | |
3561 | |
Store at 4°C in the dark. | |
IgG2b |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Common gamma Chain/IL-2 R gamma Antibody (633162), DyLight 680, Novus Biologicals™ > 0.1 mL; DyLight 680
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido