Learn More
Chitinase 3-like 1/YKL-40 Antibody (Brown U. patent anti-CHI3L1) [Janelia Fluor™ 635], Novus Biologicals™
Human Monoclonal Antibody
Marca:  Novus Biologicals (Bio-Techne) NBP3-28760JF635
Código de producto. 30159383
  • 418.00€ / 0.10 ml
Descripción
Chitinase 3-like 1/YKL-40 Monoclonal antibody specifically detects Chitinase 3-like 1/YKL-40 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Especificaciones
Especificaciones
Chitinase 3-like 1/YKL-40
Monoclonal
Janelia Fluor 635
AW208766, Brp39, Chi3l1, gp39, YKL-40
CHI3L1
0.1 mL
Cell Biology, Cytoskeleton Markers, Signal Transduction
1116
Store at 4°C in the dark.
IgG1
Flow Cytometry, ELISA, Functional Assay
Brown U. patent anti-CHI3L1
50mM Sodium Borate
Human
Protein A purified
RUO
Primary
Human
Purified
Certificados
Promociones
Promociones
Título del producto
Chitinase 3-like 1/YKL-40 Antibody (Brown U. patent anti-CHI3L1) [Janelia Fluor™ 635], Novus Biologicals™ > 0.1 mL; Janelia Fluor 635