missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ago2/eIF2C2 Antibody (11A9), Novus Biologicals™
Rat Monoclonal Antibody
Marca: Novus Biologicals NBP3-23228
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Ago2/eIF2C2 Monoclonal antibody specifically detects Ago2/eIF2C2 in Human, Bovine samples. It is validated for Western Blot
Especificaciones
Ago2/eIF2C2 | |
Monoclonal | |
Unconjugated | |
PBS | |
Rat | |
Protein G purified | |
RUO | |
Primary | |
Human, Bovine | |
Purified |
Western Blot | |
11A9 | |
Western Blot 1:250 | |
AGO2argonaute 2, argonaute2, EC 3.1.26.n1, EC 3.1.26.n2, eIF2C 2, eIF-2C 2, Eukaryotic translation initiation factor 2C 2, eukaryotic translation initiation factor 2C, 2, hAgo2, MGC3183, PAZ Piwi domain protein, PPD, protein argonaute-2, Protein slicer, Q10 | |
Clone 11A9 was generated from an Ago2/eIF2C2 synthetic peptide (coupled to KLH) corresponding to amino acids in the N-terminus of human Ago2/eIF2C2. This sequence has low homology to other argonaute family members. [UniProt# Q9UKV8] | |
0.1 mL | |
Epigenetics | |
27161 | |
Store at 4°C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG2a |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Ago2/eIF2C2 Antibody (11A9), Novus Biologicals™ > 0.1 mL; Unconjugated
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido