missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF571 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 25302-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
May be involved in transcriptional regulation.Especificaciones
ZNF571 | |
Polyclonal | |
Unconjugated | |
ZNF571 | |
HSPC059, zinc finger protein 571, ZNF571 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
51276 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q7Z3V5 | |
ZNF571 | |
ZNF571 Fusion Protein Ag18151 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |