missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF18 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 17070-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
ZNF18 be involved in transcriptional regulation.Especificaciones
ZNF18 | |
Polyclonal | |
Unconjugated | |
ZNF18 | |
HDSG1, KOX11, Zfp535, zinc finger protein 18, Zinc finger protein KOX11, ZKSCAN6, ZNF18, ZNF535 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
303226, 52712, 7566 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P17022, Q642B9, Q810A1 | |
Znf18 | |
ZNF18 Fusion Protein Ag10897 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |